Learn More
Macherey-Nagel™ MN OPTIMA™ Test Mixture in Pentane
Brand: Macherey-Nagel™ 722316
Specifications
For GC Capillary Columns | |
0.1% each of Undecane, Dodecane, Octanol, Dimethylaniline, Decylamine, Methyl Decanoate, Methyl Undecanoate, Henicosane, Docosane, Tricosane | |
1 mL | |
MN OPTIMA Test Mixture in Pentane |
Product Suggestions
Customers who viewed this item also viewed
Safety and Handling
- Test mixture for OPTIMA
Signal Word
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Aspiration toxicity Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
Supplemental information
- MIXTURE LIST-Contains : n-pentane
Your input is important to us. Please complete this form to provide feedback related to the content on this product.