Learn More
Memmert™ MobileALERT for UF75
MobileALERT provides a notification by SMS with potential free contact error option.
Brand:  Memmert™ UF75C3
Product Code. 17509655
  • 1230.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • MobileALERT
  • Notification by SMS in case of any error or alarm of the device
  • Requires option H6
Specifications
Specifications
UF75 Oven
Alerting Device
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Memmert™ MobileALERT for UF75 > For Use With: UF75