Learn More
DuPont™ Tychem 6000 F CHA5 Hooded Coverall
GREENER_CHOICE
Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.
38.45 € - 949.00 €
Specifications
Product Type Hooded Coveralls
Material Tychem
Color Gray
Waist Style Elastic
Ankle Style Elastic
View More Specs
Products 10
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11793083
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13395221
Large
928.00 €
Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
12309429
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13395221
Large
38.45 €
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
12623680
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13495186
Small
928.00 €
Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
12325659
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13395545
X-Large
38.45 €
Each
In Stock
to see stock.
Add to basket
11703093
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13395545
X-Large
928.00 €
Pack of 25
In Stock
to see stock.
Add to basket
12399419
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13395268
2X-Large
38.45 €
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
11713093
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13395268
2X-Large
928.00 €
Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
12613680
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13495156
Medium
928.00 €
Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
12319429
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13495156
Medium
38.45 €
Each
In Stock
to see stock.
Add to basket
12603680
GREENER_CHOICE
View accessory products View Documents
DuPont™
D13495118
3X-Large
949.00 €
Pack of 25
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
Description
Description
  • Available in grey for discretionary needs
  • Elasticated face, wrists, waist and ankles for optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive chin flap for tight seal of suit to face masks
  • Self-adhesive double zipper flap closure system
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
Specifications
Specifications
Hooded Coveralls
Gray
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
Self-adhesive chin flap
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
Tychem
Elastic
Elastic
Elastic
Stitched, Taped
Chemicals
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
DuPont™ Tychem 6000 F CHA5 Hooded Coverall