missing translation for 'onlineSavingsMsg'
Learn More

DuPont™ Tychem 6000 F CHA5 Hooded Coverall
GREENER_CHOICE

Offering excellent chemical permeation protection to an extensive range of chemicals. DuPont™ Tychem™ 6000 F Hooded Coveralls are made from a lightweight and durable fabric consisting of a proprietary barrier film-laminated to a heavy duty Tyvek substrate.

Brand:  DuPont™ D13395268

35.19 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 12399419

  • 38.45 € / Each
Estimated Shipment: 20-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in grey for discretionary needs
  • Elasticated face, wrists, waist and ankles for optimal fit
  • Thumb loops prevent sleeves from riding up
  • Self-adhesive chin flap for tight seal of suit to face masks
  • Self-adhesive double zipper flap closure system
  • Stitched and over-taped seams with barrier tape for protection and strength
  • Protects personnel against numerous toxic industrial organic chemicals, highly concentrated inorganic chemicals (even under pressure), particulates, radioactive contamination, biohazards and certain chemical warfare agents
  • Used for a broad range of applications from chemical spill clean-up, emergency response to military and petrochemical applications
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Hooded Coveralls
Tychem
Elastic
Elastic
Elastic
Stitched, Taped
Chemicals
(EU) 2016/425, EN 14126, EN 1073-2, EN 1149-5
For a broad range of applications from chemical spill clean-up, emergency response, to military and petrochemical applications
2X-Large
Gray
Elastic
Hooded
Double Zipper with Self-adhesive Flap
Category III, Type 3-B, 4-B, 5-B and 6-B
Self-adhesive chin flap
116 to 124 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
DuPont™ Tychem 6000 F CHA5 Hooded Coverall > Size: 2X-Large

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.