Learn More
Corning™ Molecular Biology Reagents
BIOPHARMA
Used in molecular biology, nucleic acid purification applications
Brand:  Corning™ 46-031-CM
271.98 EUR valid until 2024-12-20
Use promo code "21695" to get your promotional price.
 View more versions of this product
Product Code. 15393711
  • 286.29 € / Pack of 6
Explore more special offers
Description
Description
  • Certificate of Analysis available
  • Product can be diluted to obtain the desired concentration
Order Info
Shipped at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tris Hydrochloride Buffer
8 ±0.1
Sterile
1M Tris-Hydrochloride Buffers, 1M Tris-HCI; Liquid,; 6 x 1L
6 x 1 L