missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ Molecular Biology Reagents

Used in molecular biology, nucleic acid purification applications
263.46 € - 355.33 €
Product Code | Brand | Description | pH | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | pH | Price | Quantity & Availability | ||||
15343711
![]() |
Corning™
46-010-CM |
TAE Buffer; 10X; 6 x 1L | 8.3 ±0.1 |
263.46 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15383711
![]() |
Corning™
46-030-CM |
1M Tris-Hydrochloride Buffers, 1M Tris-HCI; 6 x 1L | 7.5 ±0.1 |
355.33 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15353711
![]() |
Corning™
46-011-CM |
TBE Buffer; 10X; 6 x 1L | 8.4 ±0.1 |
309.37 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15393711
![]() |
Corning™
46-031-CM |
1M Tris-Hydrochloride Buffers, 1M Tris-HCI; Liquid,; 6 x 1L | 8 ±0.1 |
286.29 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15313591
![]() |
Corning™
46-020-CM |
SSC Buffer 20X; 6 x 1L | 7 ±0.1 |
272.20 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Certificate of Analysis available
- Product can be diluted to obtain the desired concentration
Order Info
Shipped at ambient temperature
Safety and Handling
Recommended Storage : 15° to 30°C
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title