Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of species in enriched food and environmental samples.
Brand: 3M™ 7100041178
This item is not returnable.
View return policy
Description
- One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
- Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
- Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
- The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
- Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Listeria MDA2LIS96 | |
Microbiology Media | |
96 |
Listeria species | |
Environmental Test |