Learn More
3M™ Molecular Detection Assay 2
Used with the 3M™ Molecular Detection System for the rapid and specific detection of species in enriched food and environmental samples.
POA
Specifications
Product Type Microbiology Media
Quantity 96
View More Specs
Products 3
Product Code Brand Detects Price Quantity & Availability  
Product Code Brand Detects Price Quantity & Availability  
17298530
Encompass
missing translation for 'viewDocuments'
3M™
7100041177
Listeria monocytogenes species
POA
1 set
View alternative products
missing translation for 'toSaveUpto' 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17208540
Encompass
missing translation for 'viewDocuments'
3M™
7100041178
Listeria species
POA
1 set
View alternative products
missing translation for 'toSaveUpto' 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
17218540
Encompass
missing translation for 'viewDocuments'
3M™
7100062349
Salmonella species
POA
1 set
View alternative products
missing translation for 'toSaveUpto' 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
  • One assay protocol and fewer steps than most tests reduces repetitive tasks and makes it easy to train employees.
  • Ready-to-use and pre-dispensed reagents provide convenience and minimize the chance of human error.
  • Color-coded assay tubes by organism and matching software screens allow for better tracking and easy identification.
  • The ability to run all assays simultaneously and the flexibility to test up to 96 samples in each run helps you operate more efficiently.
  • Real-time amplification and detection process identifies positive results in as early as 15 minutes and allows you to make critical decisions faster.
Specifications
Specifications
Microbiology Media
96