missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Dexamethasone Fluorescein

The synthetic steroid hormone dexamethasone binds to the glucocorticoid receptor, producing a steroid-receptor complex that then localizes in the nucleus and regulates gene transcription.

Brand:  Invitrogen™ D1383

201.40 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11560206

  • 265.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The synthetic steroid hormone dexamethasone binds to the glucocorticoid receptor, producing a steroid-receptor complex that then localizes in the nucleus and regulates gene transcription. The green fluorescent analog, fluorescein dexamethasone should be useful for studying the mechanism of glucocorticoid receptor activation, although at a slower rate than unmodified dexamethasone.

For stock concentrations up to 1 mM, DMF or aqueous buffers above pH 6 are recommended as diluent. Stock solutions can be stored as single-use aliquots, protected from light at -20°C.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mg
Classic Dyes
Solid
494/519 nm
Room Temperature
C45H45FN2O9S2
Dexamethasone Fluorescein
840.98
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2978490 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog NumberD1383
Lot Number2850238 Certificate TypeCertificate of Analysis Date14/12/2023 Catalog NumberD1383
Lot Number2530221 Certificate TypeCertificate of Analysis Date05/10/2022 Catalog NumberD1383
Lot Number2431371 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog NumberD1383
Lot Number2349075 Certificate TypeCertificate of Analysis Date26/05/2021 Catalog NumberD1383
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Dexamethasone Fluorescein > 5 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.