missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EdU (5-ethynyl-2'-deoxyuridine)
GREENER_CHOICE

EdU can be used as a replacement for BrdU and directly measures de novo DNA synthesis or S-phase synthesis of the cell cycle using click chemistry.

Brand:  Invitrogen™ A10044

322.70 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 11590926

  • 461.00 € / 50mg

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

EdU (5-ethynyl-2’-deoxyuridine) can be used as a replacement for BrdU (5-bromo-2’-deoxyuridine) and directly measures de novo DNA synthesis or S-phase synthesis of the cell cycle using click chemistry. Click chemistry is a method of covalently coupling an azide with an alkyne. Detection of EdU employs the copper(I) catalyzed click reaction with an azide modified fluorescent dye to form a stable triazole ring. Because of the small size of the click detection reagent, no harsh denaturation steps are needed to gain access to the DNA. Eliminating this step allows for more reproducible results, a simpler and quicker protocol and measurements which can easily be multiplexed with relevant antibody based targets including phospho-histone H3, Ki-67, and cyclin B1 as well as dual-pulse experiments with BrdU by flow cytometry, fluorescence microscopy, microplate (HTS) or high-throughput imaging (HCS).

EdU is readily soluble in DMSO, alcohol, water, or aqueous buffers. For use with in vitro applications, prepare 10 mM EdU stock solution in DMSO or aqueous buffer.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

252.23 g/mol
Store in freezer (-5°C to -30°C).
Less hazardous
50 mg
EdU
Room Temperature
EdU
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3079930 Certificate TypeCertificate of Analysis Date07/11/2024 Catalog NumberA10044
Lot Number3023517 Certificate TypeCertificate of Analysis Date17/09/2024 Catalog NumberA10044
Lot Number2936396 Certificate TypeCertificate of Analysis Date01/08/2024 Catalog NumberA10044
Lot Number2907386 Certificate TypeCertificate of Analysis Date07/06/2024 Catalog NumberA10044
Lot Number2873165 Certificate TypeCertificate of Analysis Date26/03/2024 Catalog NumberA10044
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ EdU (5-ethynyl-2'-deoxyuridine) > 50 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.