missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EdU (5-ethynyl-2'-deoxyuridine)
GREENER_CHOICE

EdU can be used as a replacement for BrdU and directly measures de novo DNA synthesis or S-phase synthesis of the cell cycle using click chemistry.

456.00 € - 6340.00 €

Specifications

Shipping Condition Room Temperature
Label or Dye EdU
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11590926
GREENER_CHOICE
View Documents
Invitrogen™
A10044
50 mg
456.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
10617215
View Documents
Invitrogen™
E10187
500 mg
1630.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10839370
View Documents
Invitrogen™
E10415
5 g
6340.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

EdU (5-ethynyl-2’-deoxyuridine) can be used as a replacement for BrdU (5-bromo-2’-deoxyuridine) and directly measures de novo DNA synthesis or S-phase synthesis of the cell cycle using click chemistry. Click chemistry is a method of covalently coupling an azide with an alkyne. Detection of EdU employs the copper(I) catalyzed click reaction with an azide modified fluorescent dye to form a stable triazole ring. Because of the small size of the click detection reagent, no harsh denaturation steps are needed to gain access to the DNA. Eliminating this step allows for more reproducible results, a simpler and quicker protocol and measurements which can easily be multiplexed with relevant antibody based targets including phospho-histone H3, Ki-67, and cyclin B1 as well as dual-pulse experiments with BrdU by flow cytometry, fluorescence microscopy, microplate (HTS) or high-throughput imaging (HCS).

EdU is readily soluble in DMSO, alcohol, water, or aqueous buffers. For use with in vitro applications, prepare 10 mM EdU stock solution in DMSO or aqueous buffer.

For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.

Order Info

Shipping Condition: Room Temperature

Specifications

Specifications

Room Temperature
EdU
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.