missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 647 Streptavidin (Alexa Fluor™ 647 EGF complex)

Complexes of the dye-streptavidin molecule and biotinylated EGF

Brand:  Invitrogen™ E35351

630.80 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11568956

  • 840.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Fluorescently labeled EGF has enabled scientists to investigate receptor-membrane interactions, study receptor distribution, calculate rate constants for the interaction of EGF with its receptor, and more.

  • Biotinylated conjugates utilize biotin-XX, which contains long spacer arm designed to enhance probe affinity for EGF receptor
  • EGF molecule: 53 amino acids, MW=6,045 Da
  • Number of fluorophore molecules on each EGF molecule: 1
  • Fluorescence is typically monitored using a flow cytometer, fluorescence microscope, or fluorimeter

Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Endocytosis, Exocytosis and Phagocytosis Assays, Flow Cytometry of Cellular Processes, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer (-5 to -30°C) and protect from light.
Growth Factor
Alexa Fluor™
Wet Ice
Fluorescence
Alexa Fluor Dyes
100 μg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2805764 Certificate TypeCertificate of Analysis Date31/12/2023 Catalog NumberE35351
Lot Number2595812 Certificate TypeCertificate of Analysis Date15/03/2023 Catalog NumberE35351
Lot Number2342897 Certificate TypeCertificate of Analysis Date29/10/2021 Catalog NumberE35351
Lot Number2273631 Certificate TypeCertificate of Analysis Date17/10/2020 Catalog NumberE35351
Lot Number2131702 Certificate TypeCertificate of Analysis Date14/08/2019 Catalog NumberE35351
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Epidermal Growth Factor, Biotinylated, complexed to Alexa Fluor™ 647 Streptavidin (Alexa Fluor™ 647 EGF complex) > 100μg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.