missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PBS (Phosphate-Buffered Saline) Tablets
Phosphate-buffered saline (PBS) is a balanced salt formulation used for a variety of cellular applications.
Brand: Invitrogen™ 003002
139.08 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.
This item is not returnable.
View return policy
Description
Phosphate-buffered saline (PBS) is a balanced salt formulation used for a variety of cellular applications, such as washing cells/tissue during immunohistochemistry (IHC), immunocytochemistry (ICC); transporting cells or tissue; diluting cells for counting; and preparing reagents. PBS is formulated without calcium and magnesium. Each PBS tablet should be dissolved in 100 mL of distilled water. This buffer contains 10 mM phosphate, 150 mM sodium chloride, pH 7.3 to 7.5.NOTE: These tablets are not sterile and so should not be used for cell culture or live cells without filtering through a 0.2 μM filter.
Order Info
Shipping Condition: Room Temperature

Specifications
Store at room temperature. | |
100 Tablets | |
Room Temperature |
Phosphate Buffered Saline (PBS) | |
Phosphate Buffered Saline | |
Tablet |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Phosphate-Buffered Saline (PBS)
Version: 12/09
File size: 26.8 kB
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
78938591 | Certificate of Analysis | 31/01/2025 | 003002 |
75449554B | Certificate of Analysis | 29/01/2025 | 003002 |
75449554 | Certificate of Analysis | 29/01/2025 | 003002 |
77575691 | Certificate of Analysis | 29/01/2025 | 003002 |
77664365 | Certificate of Analysis | 29/01/2025 | 003002 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ PBS (Phosphate-Buffered Saline) Tablets > 100 tablets
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction