Learn More
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads, Thermo Scientific Chemicals
44.72 € - 426.12 €
Chemical Identifiers
CAS | 70955-01-0 |
---|---|
Molecular Formula | C20H25FN2O8 |
Molecular Weight (g/mol) | 440.42 |
MDL Number | MFCD00131613 |
InChI Key | FJUZEZRZKVMAMD-UHFFFAOYNA-N |
PubChem CID | 73906282 |
IUPAC Name | (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
SMILES | COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11377455
|
Thermo Scientific Alfa Aesar
L05466.30 |
250 g |
44.72 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11387455
|
Thermo Scientific Alfa Aesar
L05466.0B |
1000 g | N/A | N/A | N/A | |||||
11449820
|
Thermo Scientific Alfa Aesar
L05466.0I |
5000 g |
426.12 €
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads are useful in general gas drying applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Specifications
Hygroscopic | |
C20H25FN2O8 | |
Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone. | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 | |
Molecular sieves, 4A, 3 to 5mm (0.12 to 0.20 in) beads |
70955-01-0 | |
MFCD00131613 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
73906282 |
Safety and Handling
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.