missing translation for 'onlineSavingsMsg'
Learn More
Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals Product Code.: 10236940

Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals

Product Code. 10236940
25 g, Plastic Bottle
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10236940

Brand: Thermo Scientific Acros 214800250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color White to Beige
pH 9 to 12 (10% water suspension)
Packaging Plastic Bottle
Residual Water 3% max.
Water Adsorption 18% max.
Quantity 25 g
Physical Form Powder
MDL Number MFCD00131613
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Molecular sieves 4A, Powder, <50 micron

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465708 Certificate TypeCertificate of Analysis Date04/09/2024 Catalog Number214800025,214800010,214800100,214800250,214805000
Lot NumberA0429421 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog Number214800025,214800010,214800100,214800250,214805000
Lot NumberA0453754 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number214800025,214800010,214800100,214800250,214805000
Lot NumberA0447835 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number214800025,214800010,214800100,214800250,214805000
Lot NumberA0401324 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number214800025,214800010,214800100,214800250,214805000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.