Learn More
Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals
34.87 € - 316.58 €
Chemical Identifiers
CAS | 1317-33-5 |
---|---|
Molecular Formula | MoS2 |
Molecular Weight (g/mol) | 160.07 |
MDL Number | MFCD00003470 |
InChI Key | CWQXQMHSOZUFJS-UHFFFAOYSA-N |
Synonym | molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z |
PubChem CID | 14823 |
ChEBI | CHEBI:30704 |
SMILES | S=[Mo]=S |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11324787
|
Thermo Scientific Alfa Aesar
041827.14 |
25 g |
34.87 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11314787
|
Thermo Scientific Alfa Aesar
041827.22 |
100 g |
92.66 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11304787
|
Thermo Scientific Alfa Aesar
041827.36 |
500 g |
316.58 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molybdenum(IV) sulfide is used as a catalyst in the electrolysis of water. It is employed as a cocatalyst for desulfurization in petrochemistry especially hydrodesulfurization. It is also used as a component of photoelectrochemical and microelectronics applications. Further, it is useful as a filament material in many electrical applications. It finds application as a lubricant in aircraft engines. Polymers consisting of molybdenum(IV) sulfide such as nylon, teflon and vespel posses improved strength and reduced friction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-33-5 | |
160.07 | |
CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
14823 | |
S=[Mo]=S |
MoS2 | |
MFCD00003470 | |
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
CHEBI:30704 |
Specifications
1185°C | |
Black | |
99% (metals basis) | |
MFCD00003470 | |
Soluble in hot sulfuric acid,and aquaregia. Insoluble in water,concentrated sulfuric acid and dilute acid. | |
S=[Mo]=S | |
160.07 | |
CHEBI:30704 | |
Odorless | |
Molybdenum(IV) sulfide |
1317-33-5 | |
Powder | |
MoS2 | |
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
disulfanylidenemolybdenum | |
14823 | |
160.08 | |
Black powder |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves.
P261-P271-P304+P340-P312
H332
missing translation for 'einecsNumber' : 215-263-9
missing translation for 'rtecsNumber' : QA4697000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Molybdenum(IV) sulfide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
Precautionary Statement
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Molybdenum sulfide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.