missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals

144.00 € - 501.00 €

Chemical Identifiers

CAS 209733-38-0
Molecular Formula C15H35MoO5
Molecular Weight (g/mol) 391.39
MDL Number MFCD00210636
InChI Key HLFWWCCBFJUYJL-UHFFFAOYSA-N
Synonym molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
PubChem CID 18475294
IUPAC Name molybdenum;propan-2-olate
SMILES CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11374797
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041836.AC
25 mL
144.00 €
25mL
Please to purchase this item. Need a web account? Register with us today!
11364797
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041836.AE
100 mL
501.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Used in the preparations of nanomaterials and thin films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
Specifications

Chemical Identifiers

209733-38-0
391.39
HLFWWCCBFJUYJL-UHFFFAOYSA-N
18475294
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C
C15H35MoO5
MFCD00210636
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
molybdenum;propan-2-olate

Specifications

209733-38-0
12°C (54°F)
ChemSeal™ bottles
Mo[OCH(CH3)2]5
UN1219
molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
HLFWWCCBFJUYJL-UHFFFAOYSA-N
molybdenum;propan-2-olate
18475294
99.6%
Liquid
Molybdenum(V) isopropoxide
Yellow to Brown
Alcohol-like
C15H35MoO5
MFCD00210636
Moisture sensitive
Soluble in hydrocarbons,reacts with alcohols,ketones and esters
CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C
391.39
391.04
5% w/v in isopropanol
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H336
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause drowsiness or dizziness.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P370+P378q-P501c

H225-H319-H335-H336

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ISOPROPANOL

missing translation for 'tsca' : No

missing translation for 'storageNote' :

  • Packaged under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only