missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Mono Pocket Sampler

Quick and easy to use sampler that has a GMP design, which means it can be stripped and cleaned very quickly. Sampling Systems™ Mono Pocket Sampler is suitable for taking discrete, zone samples from free flowing powders and granules.

Brand:  Sampling Systems™ 1430S-600

 View more versions of this product

Product Code. 15809261

  • 400.00 € / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For sampling free flowing powders and granules
  • Chemically resistant 316L stainless steel construction
  • Easy clean design
  • Quick and easy to use
  • Simply push into your powder and granules and twist the handle to take the sample
  • Ideal for chemical, food and pharmaceutical sampling
Specifications

Specifications

Pocket Sampler
One Sampling Point
Powder Sampling
600 mm
Stainless Steel (316L)
20 mL
25 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.