Learn More
Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera
Use in slide agglutination testing for serotyping of Salmonella isolates according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera is a comprehensive range of monovalent Salmonella antigens delivered in economical 3.0mL bottles.
110.00 €
Specifications
Certifications/Compliance | W.H.O. |
---|---|
Content And Storage | 2°C to 8°C, Do Not Freeze |
Product Type | Antiserum |
For Use With (Application) | Traditional Slide Agglutination Testing |
Packaging Type | Dropper Bottle |
Product Code | Brand | Grade | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Grade | Price | Quantity & Availability | |||||
11999084
|
Pro-Lab Diagnostics™
PL.6115 |
Antigen Factor eh |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11979084
|
Pro-Lab Diagnostics™
PL.6113 |
Antigen Factor d |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11929094
|
Pro-Lab Diagnostics™
PL.6118 |
Antigen Factor h |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11959094
|
Pro-Lab Diagnostics™
PL.6122 |
Antigen Factor gm |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11989094
|
Pro-Lab Diagnostics™
PL.6125 |
Antigen Factor u |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11999094
|
Pro-Lab Diagnostics™
PL.6126 |
Antigen Factor s |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11919104
|
Pro-Lab Diagnostics™
PL.6128 |
Antigen Factor t |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11929104
|
Pro-Lab Diagnostics™
PL.6129 |
Antigen Factor f |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11939104
|
Pro-Lab Diagnostics™
PL.6131 |
Antigen Factor q |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11959104
|
Pro-Lab Diagnostics™
PL.6134 |
Antigen Factor k |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11979104
|
Pro-Lab Diagnostics™
PL.6136 |
Antigen Factor l, w |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11989104
|
Pro-Lab Diagnostics™
PL.6137 |
Antigen Factor l, v |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11919114
|
Pro-Lab Diagnostics™
PL.6140 |
Antigen Factor z13 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11989114
|
Pro-Lab Diagnostics™
PL.6147 |
Antigen Factor z24 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11999114
|
Pro-Lab Diagnostics™
PL.6148 |
Antigen Factor z32 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11919124
|
Pro-Lab Diagnostics™
PL.6151 |
Antigen Factor z29 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11929124
|
Pro-Lab Diagnostics™
PL.6153 |
Antigen Factor 2 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11939124
|
Pro-Lab Diagnostics™
PL.6154 |
Antigen Factor 5 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11969124
|
Pro-Lab Diagnostics™
PL.6157 |
Antigen Factor z6 |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11909094
|
Pro-Lab Diagnostics™
PL.6116 |
Antigen Factor enx |
110.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.
- Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
- Produced using reference strains prepared by World Health Organization guidelines
- Certified reference center performance
- Presented in color-coded dropper bottles
- Long shelf life: up to 5 years from date of manufacture
Specifications
W.H.O. | |
Antiserum | |
Dropper Bottle | |
Agglutination |
2°C to 8°C, Do Not Freeze | |
Traditional Slide Agglutination Testing | |
3.0 mL |
Safety and Handling
missing translation for 'shelfLife' : 5 Years
Your input is important to us. Please complete this form to provide feedback related to the content on this product.