Learn More
Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera
Use in slide agglutination testing for serotyping of Salmonella isolates according to Kauffmann-White classification. Pro-Lab Diagnostics™ Monovalent Flagella (H) Salmonella Antisera is a comprehensive range of monovalent Salmonella antigens delivered in economical 3.0mL bottles.
Brand: Pro-Lab Diagnostics™ PL.6113
Description
While polyvalent O (somatic) antisera aid in initial serogrouping, full identification of O antigens is achieved using monovalent-specific O antisera. The serotype of Salmonella isolates can then be determined using H (flagella) antisera.
- Ready-to-use diluted antisera with 0.01% thimerosal as a preservative
- Produced using reference strains prepared by World Health Organization guidelines
- Certified reference center performance
- Presented in color-coded dropper bottles
- Long shelf life: up to 5 years from date of manufacture
Specifications
W.H.O. | |
Antigen Factor d | |
Traditional Slide Agglutination Testing | |
3.0 mL |
2°C to 8°C, Do Not Freeze | |
Antiserum | |
Dropper Bottle | |
Agglutination |
Product Suggestions
Customers who viewed this item also viewed.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.