missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morpholine, 99.5%, purified by redistillation, AcroSeal™, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -5.0°C |
Density | 0.9900g/mL |
Boiling Point | 126.0°C to 130.0°C |
Flash Point | 32°C |
Assay Percent Range | 99.4% min. (GC) |
Packaging | AcroSeal™ Glass bottle |
Refractive Index | 1.4530 to 1.4550 |
Quantity | 100 mL |
Merck Index | 15,6362 |
Specific Gravity | 0.99 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0471886 | Certificate of Analysis | 10/04/2025 | 433851000,433858000 |
A0460380 | Certificate of Analysis | 16/02/2024 | 433851000,433858000 |
A0453790 | Certificate of Analysis | 25/08/2023 | 433851000,433858000 |
A0450334 | Certificate of Analysis | 10/02/2023 | 433851000,433858000 |
A0408926 | Certificate of Analysis | 13/12/2022 | 433851000,433858000 |
Safety and Handling



Product Identifier
- Morpholine
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 3
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H302-Harmful if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Morpholine, 99.5%, purified by redistillation, AcroSeal™, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction