missing translation for 'onlineSavingsMsg'
Learn More
Morpholine, 99%, Thermo Scientific Chemicals Product Code.: 11424153

Morpholine, 99%, Thermo Scientific Chemicals

Product Code. 11424153
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11424153

Brand: Thermo Scientific Alfa Aesar A10355.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Morpholine is a common additive for pH adjustment in both fossil fuel and nuclear power plant steam systems. It is widely used in organic synthesis as a building block in the preparation of the antibiotic linezolid, the anticancer agent gefitinib and the analgesic dextromoramide. It is also used as a solvent in chemical reactions and in the preparation of enamines. Further, it serves as a chemical emulsifier in the process of waxing fruit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Morpholine is a common additive for pH adjustment in both fossil fuel and nuclear power plant steam systems. It is widely used in organic synthesis as a building block in the preparation of the antibiotic linezolid, the anticancer agent gefitinib and the analgesic dextromoramide. It is also used as a solvent in chemical reactions and in the preparation of enamines. Further, it serves as a chemical emulsifier in the process of waxing fruit.

Solubility
Miscible with water.      

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-91-8
Molecular Formula C4H9NO
Molecular Weight (g/mol) 87.12
MDL Number MFCD00005972
InChI Key YNAVUWVOSKDBBP-UHFFFAOYSA-N
Synonym 1-oxa-4-azacyclohexane, tetrahydro-1,4-oxazine, diethylene oximide, diethylenimide oxide, diethyleneimide oxide, drewamine, diethylene imidoxide, tetrahydro-p-oxazine, tetrahydro-2h-1,4-oxazine, p-isoxazine, tetrahydroShow MoreShow Less
PubChem CID 8083
ChEBI CHEBI:34856
IUPAC Name morpholine
SMILES C1COCCN1

Specifications

Melting Point -5°C
Density 1.007
Boiling Point 129°C
Flash Point 35°C (95°F)
Assay Percent Range 99%
Odor Amine-like
Refractive Index 1.454
Quantity 100 g
UN Number UN2054
Beilstein 102549
Sensitivity Hygroscopic
Merck Index 14,6277
Solubility Information Miscible with water.
Formula Weight 87.12
Percent Purity 99%
Chemical Name or Material Morpholine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463195 Certificate TypeCertificate of Analysis Date12/11/2024 Catalog NumberA10355.36,A10355.22,A10355.0E
Lot NumberA0460121 Certificate TypeCertificate of Analysis Date08/10/2024 Catalog NumberA10355.36,A10355.22,A10355.0E
Lot NumberU06E014 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog NumberA10355.36,A10355.22,A10355.0E
Lot NumberQ18F056 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog NumberA10355.36,A10355.22,A10355.0E
Lot NumberA0456426 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog NumberA10355.36,A10355.22,A10355.0E
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Morpholine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Morpholine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Morpholine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.