missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Moulded Polyethylene Connecting Link

Used to permanently attach lengths of chain or to repair chain link, our durable B-900 Moulded Polyethylene Connecting Links match our posts and chain.

Brand:  Brady™ 078248

 View more versions of this product

Product Code. 11751296

  • 72.75 € / 1 pack
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used to permanently attach lengths of chain or to repair chain link, our durable B-900 polyethylene Connecting Links match our posts and chain.
Specifications

Specifications

36 Pk.
Black
Traffic Control
2 in.
50.8 mm
Chain Link
Polyethylene
50.8 mm
2 in.
Stanchion Accessories
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers