missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Fc gamma RIII (CD16) Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 12 publications
Brand: R&D Systems MAB19601
388.80 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
This item is not returnable.
View return policy
Description
Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
Fc gamma RIII (CD16) | |
Monoclonal | |
LYOPH | |
Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
Q5D5J5 | |
Fcgr3 | |
Mouse myeloma cell line NS0-derived recombinant mouse Fc gamma RIII Ala31-Thr215 Accession # Q5D5J5 | |
100 μg | |
Primary | |
Detects mouse Fc gamma RIIIA/B (CD16) in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Fcrl3/CD16-2, rmFc gamma RIA or rhFc gamma RIIA is observed. | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
275003 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD16, CD16A, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGRIII, FCR-10, FCRIII, FCRIIIA, IGFR3, IMD20 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
2214 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 12
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Mouse Fc gamma RIII (CD16) Antibody, R&D Systems™ > 100ug; Unlabeled
Spot an opportunity for improvement?Share a Content Correction