missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-7 (KGF) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
Brand: Gibco™ 450-60-10UG
180.42 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P36363 | |
protein with no preservative | |
18.8 | |
10 μg | |
-20°C | |
<1 EU/μg | |
FGF7 | |
Unconjugated | |
CNDMSPEQTA TSVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNSYNIMEIR TVAVGIVAIK GVESEYYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHSGG EMFVALNQKG IPVKGKKTKK EQKTAHFLPM AIT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |
Functional Assay | |
14178 | |
Mouse FGF-7 (KGF) | |
E. coli | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Mouse | |
Recombinant | |
Lyophilized |