missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Mouse FGF-7 (KGF) Recombinant Protein, PeproTech®
E. coli Recombinant Protein
80.00 € - 5140.00 €
Specifications
Accession Number | P36363 |
---|---|
For Use With (Application) | Functional Assay |
Formulation | protein with no preservative |
Gene ID (Entrez) | 14178 |
Molecular Weight (g/mol) | 18.8 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17863853
|
Gibco™
450-60-100UG |
100 μg |
1030.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17884563
|
Gibco™
450-60-10UG |
10 μg |
194.00 €
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17844123
|
Gibco™
450-60-1MG |
10 x 100 μg | N/A | ||||||
17824433
|
Gibco™
450-60-250UG |
2.5 x 100 μg | N/A | ||||||
17893983
|
Gibco™
450-60-2UG |
2 μg |
80.00 €
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17854373
|
Gibco™
450-60-500UG |
5 x 100 μg | N/A | ||||||
17883923
|
Gibco™
450-60-50UG |
50 μg |
620.00 €
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.Specifications
P36363 | |
protein with no preservative | |
18.8 | |
E. coli | |
RUO | |
Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf | |
Mouse | |
Recombinant | |
Lyophilized |
Functional Assay | |
14178 | |
Mouse FGF-7 (KGF) | |
-20°C | |
<1 EU/μg | |
FGF7 | |
Unconjugated | |
CNDMSPEQTA TSVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNSYNIMEIR TVAVGIVAIK GVESEYYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHSGG EMFVALNQKG IPVKGKKTKK EQKTAHFLPM AIT | |
≥ 95% by SDS-PAGE gel and HPLC analyses. |