Learn More
R&D Systems Mouse Luminex™ Discovery Assay
Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.
Brand:  R&D Systems LXSAMSM-12
2188.00 EUR valid until 2025-03-28
Use promo code "25306" to get your promotional price.
 View more versions of this product
Product Code. 18792214
  • 2735.00 € / 1 set
Explore more special offers
Specifications
12 Plex
Mouse
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.