missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Mouse Luminex™ Discovery Assay

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

643.00 € - 7525.00 €

Specifications

Species Mouse
Binding Target Multiplex Cytokine Assay Kits
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
View More Specs

Products 50
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18752537
View Documents
R&D Systems
LXSAMSM-04
4 Plex
1135.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30001359
View Documents
R&D Systems
LXSAMSM-01
1 Plex
643.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
30000895
View Documents
R&D Systems
LXSAMSM-03
3 Plex
929.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
18742014
View Documents
R&D Systems
LXSAMSM-17
17 Plex
3655.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18721984
View Documents
R&D Systems
LXSAMSM-14
14 Plex
3100.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18711984
View Documents
R&D Systems
LXSAMSM-06
6 Plex
1545.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18702224
View Documents
R&D Systems
LXSAMSM-19
19 Plex
4020.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18792214
View Documents
R&D Systems
LXSAMSM-12
12 Plex
2735.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18742174
View Documents
R&D Systems
LXSAMSM-09
9 Plex
2160.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18742094
View Documents
R&D Systems
LXSAMSM-15
15 Plex
3285.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18742034
View Documents
R&D Systems
LXSAMSM-24
24 Plex
4940.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18771874
View Documents
R&D Systems
LXSAMSM-21
21 Plex
4385.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18731874
View Documents
R&D Systems
LXSAMSM-05
5 Plex
1340.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18782004
View Documents
R&D Systems
LXSAMSM-31
31 Plex
5835.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18761874
View Documents
R&D Systems
LXSAMSM-16
16 Plex
3470.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18712184
View Documents
R&D Systems
LXSAMSM-07
7 Plex
1750.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18701844
View Documents
R&D Systems
LXSAMSM-32
32 Plex
5955.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18782394
View Documents
R&D Systems
LXSAMSM-11
11 Plex
2550.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18782374
View Documents
R&D Systems
LXSAMSM-22
22 Plex
4570.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18742374
View Documents
R&D Systems
LXSAMSM-18
18 Plex
3835.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18742444
View Documents
R&D Systems
LXSAMSM-27
27 Plex
5360.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18712104
View Documents
R&D Systems
LXSAMSM-02
2 Plex
724.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18762084
View Documents
R&D Systems
LXSAMSM-23
23 Plex
4755.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18781944
View Documents
R&D Systems
LXSAMSM-08
8 Plex
1955.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18712404
View Documents
R&D Systems
LXSAMSM-20
20 Plex
4205.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18762324
View Documents
R&D Systems
LXSAMSM-13
13 Plex
2920.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18732134
View Documents
R&D Systems
LXSAMSM-10
10 Plex
2370.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18432420
View Documents
R&D Systems
LXSAMSM-48
48 Plex
7365.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18432020
View Documents
R&D Systems
LXSAMSM-40
40 Plex
6715.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18442650
View Documents
R&D Systems
LXSAMSM-37
37 Plex
6470.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18472020
View Documents
R&D Systems
LXSAMSM-42
42 Plex
6875.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18411330
View Documents
R&D Systems
LXSAMSM-45
45 Plex
7120.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18432220
View Documents
R&D Systems
LXSAMSM-34
34 Plex
6190.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18491660
View Documents
R&D Systems
LXSAMSM-50
50 Plex
7525.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18471860
View Documents
R&D Systems
LXSAMSM-30
30 Plex
5715.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18432110
View Documents
R&D Systems
LXSAMSM-26
26 Plex
5240.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18402450
View Documents
R&D Systems
LXSAMSM-35
35 Plex
6310.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18482440
View Documents
R&D Systems
LXSAMSM-28
28 Plex
5480.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18451490
View Documents
R&D Systems
LXSAMSM-49
49 Plex
7445.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18491200
View Documents
R&D Systems
LXSAMSM-47
47 Plex
7280.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18402280
View Documents
R&D Systems
LXSAMSM-43
43 Plex
6960.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18482180
View Documents
R&D Systems
LXSAMSM-46
46 Plex
7200.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18492200
View Documents
R&D Systems
LXSAMSM-25
25 Plex
5120.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18462510
View Documents
R&D Systems
LXSAMSM-29
29 Plex
5595.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18481820
View Documents
R&D Systems
LXSAMSM-38
38 Plex
6555.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18412080
View Documents
R&D Systems
LXSAMSM-36
36 Plex
6390.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18451390
View Documents
R&D Systems
LXSAMSM-44
44 Plex
7040.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18472610
View Documents
R&D Systems
LXSAMSM-41
41 Plex
6795.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18491820
View Documents
R&D Systems
LXSAMSM-33
33 Plex
6070.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
18421490
View Documents
R&D Systems
LXSAMSM-39
39 Plex
6635.00 €
1 set
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

Mouse
Multiplex Cytokine Assay Kits
Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Videos
SDS
Documents

Documents

Product Certifications