Learn More
MP Biomedicals™ Casein Hydrolysate Acid
Sulfuric acid digest of casein
Brand: MP Biomedicals™ 0219102301
This item is not returnable.
View return policy
Description
Lower salt content than the hydrochloric acid digest. Essentially tryptophan free; 5.0 pH (2% soln.). Nitrogen approx. 13.0%; amino nitrogen approx. 9.2%.
Specifications
15° to 30°C | |
Amicase | |
5 to 7 | |
1 lb. |