Learn More
MP Biomedicals™ Casein Hydrolysate Acid
Sulfuric acid digest of casein
251.22 € - 439.90 €
Product Code | Brand | pH | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | pH | Quantity | Price | Quantity & Availability | ||||
11426131
|
MP Biomedicals™
0219102301 |
5 to 7 | 1 lb. |
251.22 €
1lb |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11493230
|
MP Biomedicals™
0210129105 |
5 to 7 | 5 lb. |
439.90 €
5lb |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Lower salt content than the hydrochloric acid digest. Essentially tryptophan free; 5.0 pH (2% soln.). Nitrogen approx. 13.0%; amino nitrogen approx. 9.2%.
Specifications
15° to 30°C | |
Amicase |