Learn More
MP Biomedicals™ Casein Hydrolysate Acid
Sulfuric acid digest of casein
Brand:  MP Biomedicals™ 0210129105
 View more versions of this product
Product Code. 11493230
  • 600.00 € / 5lb
Explore more special offers
Description
Description
Lower salt content than the hydrochloric acid digest. Essentially tryptophan free; 5.0 pH (2% soln.). Nitrogen approx. 13.0%; amino nitrogen approx. 9.2%.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
15° to 30°C
Amicase
5 to 7
5 lb.