missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Screw Caps

These 2 mL Screw Caps are for tubes to be used on FastPrep™ instruments or any bead beater.

Brand:  MP Biomedicals™ 115065005

349.79 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12733667

This item is not returnable. View return policy

Description

Description

  • Screw Caps with O-rings fit 2 mL screw cap tubes and are ideal for high-speed sample preparation instruments.
  • Screw Caps fit Empty Fastprep™ Tubes and are used in Fastprep systems, to hold sample for Homogenization.
  • Available separately for those who wish to fill their own Lysing Matrix Tubes.
  • Adapter Compatibility: ConeFlex Legacy (allows all FP24 adapters on FP96), CoolPrep (24x2 mL), HiPrep (48x2 mL), QuickFlex (96x2 mL), QuickPrep (24x2 mL), QuickPrep All Metal (24x2 mL)
  • Screw Caps can also be found in the FastDNA and FastDNA SPIN Kits.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2 mL
12.8 mm
0.24 in.
Blue
1000 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.