Learn More
MP Biomedicals™ Screw Caps
These 2 mL Screw Caps are for tubes to be used on FastPrep™ instruments or any bead beater.
216.00 € - 444.00 €
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Color | Price | Quantity & Availability | ||||
11472340
|
MP Biomedicals™
115066002 |
500 Pk. | Red |
216.00 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11402340
|
MP Biomedicals™
115063002 |
500 Pk. | Orange |
355.00 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11402350
|
MP Biomedicals™
115067002 |
500 Pk. | Green |
302.00 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11442340
|
MP Biomedicals™
115065002 |
500 Pk. | Blue |
333.00 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11422340
|
MP Biomedicals™
115064002 |
500 Pk. | Purple |
299.00 €
Pack of 500 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11412340
|
MP Biomedicals™
115063005 |
1000 Pk. | Orange |
377.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12733667
|
MP Biomedicals™
115065005 |
1000 Pk. | Blue |
444.00 €
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Screw Caps with O-rings fit 2 mL screw cap tubes and are ideal for high-speed sample preparation instruments.
- Screw Caps fit Empty Fastprep™ Tubes and are used in Fastprep systems, to hold sample for Homogenization.
- Available separately for those who wish to fill their own Lysing Matrix Tubes.
- Adapter Compatibility: ConeFlex Legacy (allows all FP24 adapters on FP96), CoolPrep (24x2 mL), HiPrep (48x2 mL), QuickFlex (96x2 mL), QuickPrep (24x2 mL), QuickPrep All Metal (24x2 mL)
- Screw Caps can also be found in the FastDNA and FastDNA SPIN Kits.
Specifications
2 mL |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.