missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fibrinogen from Bovine Plasma

Off-white shiny flaky powder

Brand:  MP Biomedicals™ 0215112205

 View more versions of this product

Product Code. 11496922

  • 616.92 € / 5g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Fibrinogen is blood protein that is involved in clotting and is converted to fibrin by thrombin. It consists of three non-identical pairs of disulfidebonded chains. The a-chain has an approximate molecular weight of 63.5 kDa, the b-chain 56 kDa, and the g-chain 47 kDa.

  • Molecular weight: 340kDa

Cell Biology and Analysis, Molecular biology

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Biology and Analysis, Molecular Biology
Fibrinogen from Bovine Plasma
Bovine Plasma
Powder
340
5 g
0°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers