missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fibrinogen from Bovine Plasma

Off-white shiny flaky powder

228.96 € - 2342.60 €

Specifications

For Use With (Application) Cell Biology and Analysis, Molecular Biology
Molecular Weight (g/mol) 340
Name Fibrinogen from Bovine Plasma
Source Bovine Plasma
Storage Requirements 0°C
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11496922
View Documents
MP Biomedicals™
0215112205
5 g
616.92 €
5g
Please to purchase this item. Need a web account? Register with us today!
17562716
View Documents
MP Biomedicals™
0215112201
1 g
228.96 €
1g
Please to purchase this item. Need a web account? Register with us today!
17572716
View Documents
MP Biomedicals™
0215112210
10 g
1088.62 €
10g
Please to purchase this item. Need a web account? Register with us today!
17582716
View Documents
MP Biomedicals™
0215112225
25 g
2342.60 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Fibrinogen is blood protein that is involved in clotting and is converted to fibrin by thrombin. It consists of three non-identical pairs of disulfidebonded chains. The a-chain has an approximate molecular weight of 63.5 kDa, the b-chain 56 kDa, and the g-chain 47 kDa.

  • Molecular weight: 340kDa

Cell Biology and Analysis, Molecular biology

Specifications

Specifications

Cell Biology and Analysis, Molecular Biology
Fibrinogen from Bovine Plasma
0°C
340
Bovine Plasma
Powder
Videos
SDS
Documents

Documents

Product Certifications