missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Lysozyme from Chicken Egg White

2X crystallized, salt-free and soluble

Brand:  MP Biomedicals™ 0210083405


 View more versions of this product

Product Code. 11374029

This item is not returnable. View return policy

Description

Description

Activity: >9000 units/mg protein. Unit Definition: One unit will cause a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate.

  • Lyophilized
  • Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
  • CAS: 9001-63-2

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2X
11
0°C
5 g
14388
Crystallized, Lyophilized
White
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen
Chicken egg white
Lysozyme
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Lysozyme

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P321-Specific treatment (see on this label).
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

Research Use Only