missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Lysozyme from Chicken Egg White
2X crystallized, salt-free and soluble
61.85 € - 189.00 €
Specifications
Concentration | 2X |
---|---|
Color | White |
pH | 11 |
For Use With (Application) | Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen |
Content And Storage | 0°C |
Description
Activity: >9000 units/mg protein. Unit Definition: One unit will cause a decrease in A450 of 0.001 per minute at pH 6.24 and 25°C using Micrococcus lysodeikticus as substrate.
- Lyophilized
- Synonyms: Muramidase, Mucopeptide N-acetylmuramoylhydrolase
- CAS: 9001-63-2
Specifications
2X | |
11 | |
0°C | |
Lysozyme | |
Surface-active reagents such as Dodecyl Sulfate, Alcohols and Fatty Acids, Imidazole and Indole derivatives are inhibitors via formation of change-transfer complexes |
White | |
Lysing E. coli and Streptomycetes for extraction purposes such as extracting group specific Antigen | |
Chicken egg white | |
14388 | |
Crystallized, Lyophilized |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Lysozyme
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin sensitiser Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P321-Specific treatment (see on this label).
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P501-Dispose of contents/container to in accordance with all local, regional,national and international regulations.
Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title