missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ PlantCon™ Plant Tissue Culture Container
Sterilized Base for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.
Brand: MP Biomedicals™ 092672202
79.26 EUR valid until 2024-12-31
Use promo code "21928" to get your promotional price.
This item is not returnable.
View return policy
Description
- Clear polyvinyl chloride (PVC) dishes specifically designed for plant culture applications.
- Ideal for use in commercial horticulture or research settings.
- Allow exceptional light transmission and optimal growth.
- Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization.
- Covers Sold Separately (See 092672102)
Order Info
Base L x W x H: 3 3/4 x 3 3/4 x 1 3/8 in. (9.5 x 9.5 x 3.5cm). Covers and bases are sold separately. Sterile.
Specifications
300 mL | |
Square | |
PVC | |
Cell Culture Dish | |
10/Cs | |
9.5 x 9.5 x 3.5 cm (Container), 9.5 x 9.5 x 7 cm (Cover) | |
9.5 cm |
Not Autoclavable | |
Single-use | |
Sterile | |
Plant Cell Culture | |
3.75 x 3.75 in. (9.5 x 9.5 cm) | |
9.5 cm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction