missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ PlantCon™ Plant Tissue Culture Container

Sterilized Base for PlantCon™ Plant Tissue Culture Container. Use PlantCon™ containers with liquid or agar solidified media. They can be used for many plant cell culture procedures that currently utilize glass containers.

48.55 € - 795.00 €

Specifications

Bottom Shape Square
Disposable Single-use
Material PVC
Sterility Sterile
Type Cell Culture Dish
View More Specs

Products 3
Product Code Brand Type Quantity Price Quantity & Availability  
Product Code Brand Type Quantity Price Quantity & Availability  
11415005
View Documents
MP Biomedicals™
092672002
Cell Culture Dish 1 Ea
48.55 €
Pack of 10
Estimated Shipment: 13-01-2025
to see stock.
Add to basket
 
17345609
View Documents
MP Biomedicals™
092672202
Cell Culture Dish 10/Cs
86.95 €
Pack of 10
Estimated Shipment: 10-02-2025
to see stock.
Add to basket
 
11435005
View Documents
MP Biomedicals™
092672206
Cell Culture Dish 200/Cs
795.00 €
Pack of 200
Estimated Shipment: 12-05-2025
to see stock.
Add to basket
 
Description

Description

  • Clear polyvinyl chloride (PVC) dishes specifically designed for plant culture applications.
  • Ideal for use in commercial horticulture or research settings.
  • Allow exceptional light transmission and optimal growth.
  • Reduce the time spent preparing an in vitro growth environment for plant cultures by eliminating the need for cleaning and sterilization.
  • Covers are sold separately (see 092672102)

Order Info

Base L x W x H: 3 3/4 x 3 3/4 x 1 3/8 in. (9.5 x 9.5 x 3.5cm). Covers and bases are sold separately. Sterile.

Specifications

Specifications

Square
PVC
Cell Culture Dish
Single-use
Sterile
3.75 x 3.75 in. (9.5 x 9.5 cm)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.