Learn More
MP Biomedicals™ Yeast RPM Plasmid Isolation Kit
Yeast Rapid Pure Miniprep Plasmid Isolation Kit designed for yeast cells. Can isolate plasmid from bacterial transformants for use in any downstream application (PCR, sequencing, restriction enzyme digestion, labeling, etc).
Brand:  MP Biomedicals™ 112069400
Product Code. 11363149
  • 618.00 € / 100 preps
Explore more special offers
Description
Description
  • The plasmid isolated from yeast is suitable for PCR and bacterial transformation.
  • The same kit can be used to isolate plasmid from bacterial transformants for use in any downstream application (PCR, sequencing, restriction enzyme digestion, labeling, etc).
  • Plasmid DNA mini-preps from yeast rescue positive prey plasmids after a yeast two-hybrid or a yeast one-hybrid screen.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Yeast Plasmid Prep
Yeast lysing matrix, Alkaline Lysis solution, neutralizing solution, spin Buffer, wash solution concentrate, spin filters, catch tubes, Pre-lysis Buffer
1.5 mL
For PCR and bacterial transformation
100 Preps
Yeast Culture
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.