missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Yeast, Torula, Powder

Source: Candida utilis

Brand:  MP Biomedicals™ 0290308505

 View more versions of this product

Product Code. 11405712

  • 265.00 € / 5lb

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Applications:
Torula yeast is a common flavoring additive in processed foods such as chips, seasoning blends, and crackers. It is also used in pet foods, in beekeeping, and as an attractant in traps designed to control olive flies

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Animal Diet
Torula Yeast is a common flavoring additive in processed foods such as chips, seasoning blends and crackers, also used in pet foods, in beekeeping and as an attractant in traps designed to control olive flies
15°C to 30°C
Powder
2.26 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
MP Biomedicals™ Yeast, Torula, Powder > Yeast, Torula, 5 lb.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.