Learn More
MP Biomedical™ Rapid 2019-nCoV IgG/IgM Combo Diagnostic Test Cards to COVID-19
Rapid 2019-nCoV IgG/IgM Combo Diagnostic Test Cards to COVID-19 is a rapid immunochromatographic test designed for the qualitative determination of IgG and IgM antibodies to 2019 novel coronavirus (2019-nCoV, SARS-CoV-2) in human serum, plasma, or whole blood.
Brand: MP Biomedical™ 07RDCOV19
Description
- A rapid diagnostic test for the detection of IgG and IgM antibodies to COVID-19.
- Rapid results in 15 minutes, and requires no instruments.
- It is widely accepted that IgM provides the first line of defense during viral infections, followed by the generation of adaptive, high affinity IgG responses for long term immunity and immunological memory. Testing of COVID-19 IgM and IgG antibodies therefore, is an effective method for the rapid diagnosis of COVID-19 infection. Detection of COVID-19 IgM antibodies tend to indicate a recent exposure to COVID-19, whereas detection of COVID-19 IgG antibodies indicates a later stage of infection. Thus, this combined antibody test from MP Biomedicals could also provide information on the stage of infection.
Specifications
96.8% | |
CE-IVD | |
Immunochromatography | |
87.8% | |
MP Rapid 2019-nCoV IgG/IgM Combo Test Card (x25), Sample buffer, Capillary pipets, Instructions for Use | |
25 Tests | |
2°C to 30°C | |
15 min. |
Store the test device at 2 to 30°C in the original sealed pouch. Do Not Freeze. | |
COVID-19 IgG, IgM | |
Serum, Plasma, Whole Blood | |
Single-use | |
1 Box of 25 Tests | |
2 μL | |
0.99 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.