missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MST1R (RON) Recombinant Human Protein

MST1R (RON) is a receptor tyrosine kinase.

Brand:  Invitrogen™ PV4332

 View more versions of this product

Product Code. 10026742

  • 7890.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MST1R (RON) is involved in cell proliferation and motility.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NP_002438
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
MST1R (RON)
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
74.8kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Cytoplasmic
MET (HGFR) Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers