missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MST1R (RON) Recombinant Human Protein
MST1R (RON) is a receptor tyrosine kinase.
1826.00 € - 7890.00 €
Specifications
Accession Number | NP_002438 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol |
Molecular Weight (g/mol) | 74.8kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15468633
|
Invitrogen™
PR7906A |
100μg |
1826.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10026742
|
Invitrogen™
PV4332 |
1mg |
7890.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MST1R (RON) is involved in cell proliferation and motility.
Specifications
NP_002438 | |
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Cytoplasmic | |
MET (HGFR) Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
74.8kDa | |
RUO - research use only | |
MST1R (RON) | |
Human | |
GST Tag | |
Frozen |