missing translation for 'onlineSavingsMsg'
Learn More

Genlab Multi Purpose Benchtop Ovens

Highly efficient, reliable, and cost effective ovens that suit most warming, drying, and heat treatment processes. Available in a variety of sizes and equipped with intuitive touchscreen control that offers excellent accuracies.

Brand:  Genlab MINO/40/TDIG/F

 View more versions of this product

Product Code. 16315598

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Included Features

  • On screen historical trending (48 Hours)
  • Manual or automatic overheat reset
  • High accuracy Pt100B duplex sensors
  • Excellent stability (fan assisted)
  • Low chamber uniformity
  • Energy efficient with integral energy meter
  • New - Set point lock feature

Available Options (Upon Request)

  • Fan assisted circulation (Not available < 30L)
  • Stainless steel chamber
  • 8 Stage profile control
  • Count down timer with 24/7 real time clock operation
  • 10 Menu x 8 Stage Profile Controller with Cyclic feature
  • Access ports (25, 50, 75 or 100 mm)
  • Audible warnings
  • Traceable calibration certificate to National Standards
  • Bespoke stands and stacking kits
  • Extractor unit
  • Extended warranty
  • Explosion relief panels (OV style only)
  • Bespoke solutions available upon request
Specifications

Specifications

Stainless Steel
Multi-purpose benchtop oven with base mounted controller, fan assisted
40°C to 250°C
2 Shelves, 3 Positions
<±0.6°C (Fan Assisted)
50 x 63 x 54 cm
750 W
Base Mounted, Touchscreen
40 L
Coated Steel
Chrome Plated Wire Grid Shelves
36 x 31 x 32 cm
35 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers