missing translation for 'onlineSavingsMsg'
Learn More

Buerkle™ Stainless Steel Multi-Sampler

Buerkle™ Multi Sampler features several closed chambers, for multi point sampling from several predetermined depths. The Zone Samplers are available in three different lengths, 55 cm (especially suitable for sacks), 85cm (for barrels, tubs, mixed containers) and 150 cm (for big bags, silos, tanks).

610.00 € - 1240.00 €

Specifications

Product Type Multi Sampler
Autoclavable Autoclavable
Handle Type T-shape
Certifications/Compliance DIN
For Use With (Application) For Multi Point Sampling From Several Predetermined Sampling Depths, Foodstuffs
View More Specs

Products 6
Product Code Brand Includes Length (Metric) Material Size Price Quantity & Availability  
Product Code Brand Includes Length (Metric) Material Size Price Quantity & Availability  
10255182
View accessory products View Documents Promotion Details
Buerkle™
5316-3055
3 chambers 550 mm Stainless Steel (V4A) 430 mm Insertion Depth
913.00 €
Each
In Stock
to see stock.
Add to basket
 
10268320
View accessory products View Documents Promotion Details
Buerkle™
5316-0055
3 Collection Chambers 550 mm Stainless Steel (V4A), PTFE 430 mm Insertion Depth
610.00 €
Each
In Stock
to see stock.
Add to basket
 
10265182
View accessory products View Documents Promotion Details
Buerkle™
5316-3085
3 chambers 850 mm Stainless Steel (V4A) 710 mm Insertion Depth
974.00 €
Each
In Stock
to see stock.
Add to basket
 
10024190
View accessory products View Documents Promotion Details
Buerkle™
5316-0085
3 chambers 850 mm Stainless Steel (V4A), PTFE 710 mm Insertion Depth
682.00 €
Each
In Stock
to see stock.
Add to basket
 
10192602
View accessory products View Documents Promotion Details
Buerkle™
5316-3150
5 chambers 1500 mm Stainless Steel (V4A) 1355 mm Insertion Depth
1240.00 €
Each
In Stock
to see stock.
Add to basket
 
10034190
View accessory products View Documents Promotion Details
Buerkle™
5316-0150
5 Collection Chambers 1500 mm Stainless Steel (V4A), PTFE 1355 mm Insertion Depth
995.00 €
Each
In Stock
to see stock.
Add to basket
 
Description

Description

  • All-layer samples can be collected from all layers of the container
  • Various bulk goods can be collected with the lances, from the finest powder to large grains such as corn or nuts
  • Safe and reliable sampling is made even easier with useful special accessories such as sealing sleeves, Quali-Paper, cleaning brushes, emptying devices and close-it control seals
  • aluminum, AISI 316L (1.4404) or AISI 316L/PTFE
  • Version made of AISI 316L (1.4404)/PTFE with low weight due to PTFE sample chambers.
  • With colour coding system
  • 25 mm Ø
  • Smallest possible gap between the inner and the outer tube
  • Easy cleaning, removable tip
  • Colour coding (Multi and Uno sampler)
  • Ultra-pure materials
  • Excellent processing, robust model
Specifications

Specifications

Multi Sampler
T-shape
For Multi Point Sampling From Several Predetermined Sampling Depths, Foodstuffs
Autoclavable
DIN
Videos
SDS
Documents

Documents

Certificates