missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.
394.00 € - 5745.00 €
Specifications
Autoclavable | Autoclavable (50 times at 121°C, 1 bar, 20 min.) |
---|---|
Material | Polypropylene Tip, Silicone Tubing |
Volume (Metric) | 5 to 2500 μL |
For Use With (Equipment) | Multidrop Combi, Multidrop 384, Multidrop DW |
Diameter (Metric) | Tip: 0.5 mm; Tubing: 1.3 mm |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
11799478
|
Thermo Scientific™
24070290 |
Tubing set for standard dispensing cassette (tubing length 40cm) |
394.00 €
Each |
In Stock
Log in to see stock. ![]() |
||||||
11560745
|
Thermo Scientific™
24070297 |
Long tubing set for standard dispensing cassette (tubing length 200 cm) |
652.00 €
Each |
Estimated Shipment: 30-04-2025
Log in to see stock. |
||||||
10004442
![]() |
Thermo Scientific™
24072670 |
Standard tube dispensing cassette |
740.33 €
Each |
In Stock
Log in to see stock. |
||||||
10419773
|
Thermo Scientific™
24072671 |
Standard tube dispensing cassette 5-pack |
4025.00 €
Pack of 5 |
In Stock
Log in to see stock. |
||||||
10143833
|
Thermo Scientific™
24072672 |
Standard tube dispensing cassette 10-pack |
5745.00 €
Pack of 10 |
In Stock
Log in to see stock. |
||||||
10410144
|
Thermo Scientific™
24072677 |
Long standard tube dispensing cassette (tubing length 200cm) |
1068.58 €
Each |
In Stock
Log in to see stock. |
||||||
Description
Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.
Ordering Information:
Cat Nos 24070297 and 24070290 include tip combs.
Specifications
Autoclavable (50 times at 121°C, 1 bar, 20 min.) | |
5 to 2500 μL | |
Tip: 0.5 mm; Tubing: 1.3 mm |
Polypropylene Tip, Silicone Tubing | |
Multidrop Combi, Multidrop 384, Multidrop DW |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories