missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Combi Reagent Dispenser

Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.

Brand:  Thermo Scientific™ 5840330

22194.07 EUR valid until 2024-11-29
Use promo code "21683" to get your promotional price.


 View more versions of this product

Product Code. 17801692

  • 30525.00 € / Each
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.

 The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.
  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range

Multidrop Combi SMART:

  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability

Recommended for:

  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Small tube dispensing cassette: 2μL, ±10%; 10μL, ±5% Standard tube dispensing cassette: 20μL, ±2%; 100μL, ±1%
Multidrop™ Combi+ Reagent Dispenser
0.5 to 2500 μL
Can be used with automated systems (e.g. Thermo Scientific™ Orbitor™ Microplate Mover) and a variety of microplates and strips in 96, 384 or 1536-well plate format
On-board software and Serial RS-232 or USB for PC control
  • Small tube dispensing cassette: 2 μL: CV ≤ 5%; 10 μL: CV ≤ 3% 
  • Standard tube dispensing cassette:  20 μL: CV ≤ 1.5%; 100 μL: ≤ CV 1% 
  • Small tube dispensing cassette:  0.5-50μL, in 0.5μL increment 
  • Standard tube dispensing cassette:  5-2500μL, in 5 μL increment 
1 year
20.1 lb 
Reagent Dispenser
8
The instrument is intended for automated dispensing of up to eight different reagents simultaneously into 6, 12, 24, 48, 96, or 384-well plate format. It can be used in several applications, for example, in drug discovery/high-throughput screening, genomic, proteomic and cell-based assays, and ELISA. It is capable of dispensing into 2D tubes in SLAS format that are shorter than 50mm.  
 
One standard tube dispensing cassette (cat. no. 24072670)
6-1536 - 6, 12, 24, 48, 96, and 384 well plates
  • 96-well plate: 10μL in 5 sec.; 20μL in 5sec.; 100μL in 10 sec 
  • 384-well plate: 1μL in 6 sec.; 5μL in 9 sec.; 10μL in 13 sec.; 20μL in 21 sec. 
  • 1536-well plate: 1μL in 15 sec.; 5μL in 28 sec. 
<1mL volume range 0.5-50μL; <7mL volume range >50μL (it is possible to blackflush the reagent into original reservoir to avoid loss of expensive reagent)
14 x 8.6 x 12.9 in.; 35.5 x 22 x 33 cm
9.1 kg 
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers