Learn More
ZVG Zellstoff Vertrieb™ Multitex Soft Wipes
Use ZVG Zellstoff Vertrieb™ Multitex Soft Wipes in sensitive work areas such as precision engineering, electrical and automotive industries and paint shops.
Brand:  ZVG Zellstoff Vertrieb™ 10801-01
 View more versions of this product
Product Code. 10263532
  • 107.00 € / Pack of 400
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
These high-quality nonwoven fabrics are suitable for both wet and dry cleaning due to their high strength. They are frequently used in sensitive work areas such as precision engineering, paint shops, the electrical and automotive industries, as well as in nursing homes.
  • Viscose nonwoven fabric with textile characteristics
  • Cpable of absorbing large quantities of liquid
  • Can be washed and wrung out during cleaning
  • Very soft, yet lint-free
  • Highly absorbent
  • Tear- and solvent-resistant
Viscose fibers are industrially produced from cellulose, a basic material. The chemical composition of this 'natural synthetic fiber' is similar to that of cotton. Viscose products are highly absorbent, flexible, and durable due to their textile nature.
Specifications
Specifications
Wipers
38 cm
38 x 34 cm
34 cm
White
For wet and dry cleaning