missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid Cell Marker Antibody (BM-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-32880-0.02mg
This item is not returnable.
View return policy
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Myeloid Cell Marker | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein | |
Human peripheral blood mononuclear cells | |
0.02 mg | |
Primary | |
Store at 4C. | |
IgG1 κ |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
BM-1 | |
Unconjugated | |
10 mM PBS with 0.05% BSA | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction